Assay Details
| Gene Symbol: ttl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113677.1 | NP_001107149.1 | 4 - 5 | 832 | 77 | |
| GenBank mRNA | BC157256.1 | - | 4 - 5 | 832 | 77 | |
| BC170984.1 | - | 4 - 5 | 694 | 77 | ||
| BC170986.1 | - | 4 - 5 | 694 | 77 | ||
More Information
Set Membership: |
|

ttl
Genomic Map