Assay Details
| Gene Symbol: ltb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113691.1 | NP_001107163.1 | 1 - 2 | 142 | 99 | |
| GenBank mRNA | AM041995.1 | - | 1 - 2 | 142 | 99 | |
More Information
Set Membership: |
|

ltb
Genomic Map