Assay Details
| Gene Symbol: il4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113807.1 | NP_001107279.1 | 2 - 3 | 247 | 79 | |
| GenBank mRNA | AM041996.1 | - | 2 - 3 | 247 | 79 | |
More Information
Set Membership: |
|

il4
Genomic Map