Assay Details
| Gene Symbol: dusp28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113811.1 | NP_001107283.1 | 2 - 3 | 441 | 95 | |
| XM_012970016.2 | XP_012825470.1 | 2 - 3 | 674 | 95 | ||
| XM_018093772.1 | XP_017949261.1 | 2 - 3 | 444 | 95 | ||
| GenBank mRNA | BC155536.1 | - | 2 - 3 | 441 | 95 | |
| BC171183.1 | - | 1 - 2 | 401 | 95 | ||
| BC171187.1 | - | 1 - 2 | 401 | 95 | ||
More Information
Set Membership: |
|

dusp28
Genomic Map