Assay Details
| Gene Symbol: dab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113818.1 | NP_001107290.1 | 10 - 11 | 1492 | 81 | |
| XM_018092907.1 | XP_017948396.1 | 14 - 15 | 1782 | 81 | ||
| XM_018092908.1 | XP_017948397.1 | 14 - 15 | 1782 | 81 | ||
| XM_018092909.1 | XP_017948398.1 | 13 - 14 | 1734 | 81 | ||
| XM_018092910.1 | XP_017948399.1 | 14 - 15 | 1689 | 81 | ||
| XM_018092911.1 | XP_017948400.1 | 13 - 14 | 1641 | 81 | ||
| GenBank mRNA | BC155684.1 | - | 10 - 11 | 1492 | 81 | |
| BC171185.1 | - | 11 - 12 | 1400 | 81 | ||
More Information
Set Membership: |
|

dab1
Genomic Map