Assay Details
| Gene Symbol: iqsec1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113832.1 | NP_001107304.1 | 2324 | 53 | ||
| GenBank mRNA | BC155699.1 | - | 2324 | 53 | ||
| BC171090.1 | - | 2142 | 53 | |||
More Information
Set Membership: |
|

iqsec1
Genomic Map