Assay Details
| Gene Symbol: me3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113837.1 | NP_001107309.1 | 11 - 12 | 1455 | 66 | |
| GenBank mRNA | BC155705.1 | - | 11 - 12 | 1455 | 66 | |
More Information
Set Membership: |
|

me3
Genomic Map