Assay Details
| Gene Symbol: rpl37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113853.1 | NP_001107325.1 | 2 - 3 | 229 | 64 | |
| XM_012953424.1 | XP_012808878.1 | 4 - 5 | 737 | 64 | ||
| GenBank mRNA | BC156002.1 | - | 2 - 3 | 229 | 64 | |
| CU075433.1 | - | 2 - 3 | 225 | 64 | ||
More Information
Set Membership: |
|

rpl37
Genomic Map