Assay Details
| Gene Symbol: rps7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113856.1 | NP_001107328.1 | 2 - 3 | 119 | 77 | |
| XM_012962679.2 | XP_012818133.1 | 2 - 3 | 177 | 77 | ||
| XM_012962680.2 | XP_012818134.1 | 2 - 3 | 255 | 77 | ||
| XM_012962681.2 | XP_012818135.1 | 2 - 3 | 255 | 77 | ||
| GenBank mRNA | BC077028.1 | - | 2 - 3 | 116 | 77 | |
| BC156016.1 | - | 2 - 3 | 119 | 77 | ||
| BC171191.1 | - | 2 - 3 | 117 | 77 | ||
| BC171193.1 | - | 2 - 3 | 117 | 77 | ||
| CT025297.2 | - | 1 - 2 | 93 | 77 | ||
More Information
Set Membership: |
|

rps7
Genomic Map