Assay Details
| Gene Symbol: rps7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113856.1 | NP_001107328.1 | 5 - 6 | 403 | 101 | |
| XM_012962679.2 | XP_012818133.1 | 5 - 6 | 461 | 101 | ||
| XM_012962680.2 | XP_012818134.1 | 5 - 6 | 539 | 101 | ||
| XM_012962681.2 | XP_012818135.1 | 5 - 6 | 539 | 101 | ||
| GenBank mRNA | BC077028.1 | - | 5 - 6 | 400 | 101 | |
| BC156016.1 | - | 5 - 6 | 403 | 101 | ||
| BC171191.1 | - | 5 - 6 | 401 | 101 | ||
| BC171193.1 | - | 5 - 6 | 401 | 101 | ||
| CT025297.2 | - | 4 - 5 | 377 | 101 | ||
More Information
Set Membership: |
|

rps7
Genomic Map