Assay Details
| Gene Symbol: timp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113857.1 | NP_001107329.1 | 151 | 85 | ||
| GenBank mRNA | BC156017.1 | - | 151 | 85 | ||
| BC171245.1 | - | 140 | 85 | |||
| BC171249.1 | - | 140 | 85 | |||
More Information
Set Membership: |
|

timp-1
Genomic Map