Assay Details
Gene Symbol: ccr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113862.1 | NP_001107334.1 | 1 | 442 | 84 | |
GenBank mRNA | BC155500.1 | - | 1 | 442 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
