Assay Details
| Gene Symbol: fam92a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113863.1 | NP_001107335.1 | 4 - 5 | 514 | 71 | |
| XM_012964249.1 | XP_012819703.1 | 4 - 5 | 529 | 71 | ||
| XM_012964250.1 | XP_012819704.1 | 4 - 5 | 523 | 71 | ||
| GenBank mRNA | BC157135.1 | - | 4 - 5 | 514 | 71 | |
| BC167870.1 | - | 4 - 5 | 514 | 71 | ||
| BC171287.1 | - | 4 - 5 | 458 | 71 | ||
| BC171289.1 | - | 4 - 5 | 458 | 71 | ||
More Information
Set Membership: |
|

fam92a
Genomic Map