Assay Details
| Gene Symbol: pura | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113869.1 | NP_001107341.1 | 1 | 850 | 67 | |
| GenBank mRNA | BC157149.1 | - | 1 | 850 | 67 | |
| BC171170.1 | - | 1 | 826 | 67 | ||
| BC171172.1 | - | 1 | 826 | 67 | ||
| CU025135.1 | - | 1 | 810 | 67 | ||
More Information
Set Membership: |
|

pura
Genomic Map