Assay Details
| Gene Symbol: tmem144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113871.2 | NP_001107343.2 | 9 - 10 | 888 | 64 | |
| XM_012953656.2 | XP_012809110.1 | 9 - 10 | 969 | 64 | ||
| GenBank mRNA | BC161594.1 | - | 9 - 10 | 888 | 64 | |
More Information
Set Membership: |
|

tmem144
Genomic Map