Assay Details
| Gene Symbol: rdh16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113887.1 | NP_001107359.1 | 2 - 3 | 945 | 103 | |
| XM_012956390.2 | XP_012811844.1 | 3 - 4 | 1396 | 103 | ||
| GenBank mRNA | BC157206.1 | - | 2 - 3 | 945 | 103 | |
| BC160521.1 | - | 2 - 3 | 945 | 103 | ||
| BC161248.1 | - | 3 - 4 | 649 | 103 | ||
| BC171042.1 | - | 2 - 3 | 760 | 103 | ||
| BC171044.1 | - | 2 - 3 | 760 | 103 | ||
More Information
Set Membership: |
|

rdh16
Genomic Map