Assay Details
| Gene Symbol: ap5m1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113909.1 | NP_001107381.1 | 7 - 8 | 1425 | 90 | |
| XM_012967963.2 | XP_012823417.1 | 7 - 8 | 1569 | 90 | ||
| GenBank mRNA | BC157267.1 | - | 7 - 8 | 1425 | 90 | |
| BC171155.1 | - | 6 - 7 | 1303 | 90 | ||
| CT027867.2 | - | 6 - 7 | 1345 | 90 | ||
More Information
Set Membership: |
|

ap5m1
Genomic Map