Assay Details
| Gene Symbol: neil2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113910.1 | NP_001107382.1 | 3 - 4 | 484 | 77 | |
| GenBank mRNA | BC157269.1 | - | 3 - 4 | 484 | 77 | |
More Information
Set Membership: |
|

neil2
Genomic Map