Assay Details
| Gene Symbol: chic2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113911.1 | NP_001107383.1 | 1 - 2 | 165 | 70 | |
| GenBank mRNA | BC157275.1 | - | 1 - 2 | 165 | 70 | |
| BC170969.1 | - | 1 - 2 | 140 | 70 | ||
| BC171299.1 | - | 1 - 2 | 140 | 70 | ||
More Information
Set Membership: |
|

chic2
Genomic Map