Assay Details
| Gene Symbol: wbp2nl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113925.1 | NP_001107397.1 | 7 - 8 | 830 | 97 | |
| GenBank mRNA | BC157344.1 | - | 7 - 8 | 830 | 97 | |
| BC158898.1 | - | 7 - 8 | 830 | 97 | ||
| CT027883.2 | - | 7 - 8 | 829 | 97 | ||
More Information
Set Membership: |
|

wbp2nl
Genomic Map