Assay Details
| Gene Symbol: dhrs7c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113951.1 | NP_001107423.1 | 5 - 6 | 623 | 61 | |
| XM_012971521.2 | XP_012826975.1 | 5 - 6 | 714 | 61 | ||
| GenBank mRNA | BC157553.1 | - | 5 - 6 | 623 | 61 | |
More Information
Set Membership: |
|

dhrs7c
Genomic Map