Assay Details
| Gene Symbol: myzap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113953.3 | NP_001107425.3 | 7 - 8 | 1180 | 90 | |
| GenBank mRNA | BC157558.1 | - | 7 - 8 | 778 | 90 | |
More Information
Set Membership: |
|

myzap
Genomic Map