Assay Details
| Gene Symbol: gosr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114065.1 | NP_001107537.1 | 3 - 4 | 294 | 90 | |
| XM_018097669.1 | XP_017953158.1 | 3 - 4 | 332 | 90 | ||
| GenBank mRNA | BC157789.1 | - | 3 - 4 | 294 | 90 | |
| BC170946.1 | - | 3 - 4 | 236 | 90 | ||
| BC171296.1 | - | 3 - 4 | 236 | 90 | ||
More Information
Set Membership: |
|

gosr2
Genomic Map