Assay Details
| Gene Symbol: cpa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114209.1 | NP_001107681.1 | 5 - 6 | 498 | 76 | |
| GenBank mRNA | BC088897.1 | - | 5 - 6 | 485 | 76 | |
| BC157306.2 | - | 5 - 6 | 498 | 76 | ||
More Information
Set Membership: |
|

cpa1
Genomic Map