Assay Details
| Gene Symbol: tmem120a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114210.1 | NP_001107682.1 | 10 - 11 | 878 | 74 | |
| XM_012956883.2 | XP_012812337.1 | 9 - 10 | 801 | 74 | ||
| XM_012956884.2 | XP_012812338.1 | 9 - 10 | 795 | 74 | ||
| XM_018091218.1 | XP_017946707.1 | 9 - 10 | 774 | 74 | ||
| GenBank mRNA | BC089189.1 | - | 10 - 11 | 829 | 74 | |
| BC153698.1 | - | 10 - 11 | 878 | 74 | ||
More Information
Set Membership: |
|

tmem120a
Genomic Map