Assay Details
Gene Symbol: c6orf203 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114216.1 | NP_001107688.1 | 2 - 3 | 496 | 105 | |
XM_012963124.2 | XP_012818578.1 | 2 - 3 | 531 | 105 | ||
XM_012963125.2 | XP_012818579.1 | 2 - 3 | 521 | 105 | ||
XM_012963126.2 | XP_012818580.1 | 2 - 3 | 512 | 105 | ||
XM_018093935.1 | XP_017949424.1 | 2 - 3 | 1400 | 105 | ||
XM_018093936.1 | XP_017949425.1 | 2 - 3 | 495 | 105 | ||
GenBank mRNA | BC121588.1 | - | 1 - 2 | 408 | 105 | |
BC157523.1 | - | 2 - 3 | 496 | 105 | ||
CU025092.1 | - | 2 - 3 | 445 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
