Assay Details
| Gene Symbol: pmf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114230.1 | NP_001107702.1 | 3 - 4 | 378 | 66 | |
| GenBank mRNA | BC135163.1 | - | 3 - 4 | 346 | 66 | |
| BC157753.1 | - | 3 - 4 | 378 | 66 | ||
More Information
Set Membership: |
|

pmf1
Genomic Map