Assay Details
| Gene Symbol: nrg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114241.1 | NP_001107713.1 | 1 - 2 | 1114 | 66 | |
| GenBank mRNA | BC136110.1 | - | 1 - 2 | 893 | 66 | |
| BC153324.1 | - | 1 - 2 | 1114 | 66 | ||
More Information
Set Membership: |
|

nrg3
Genomic Map