Assay Details
| Gene Symbol: cxcl14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114242.1 | NP_001107714.1 | 2 - 3 | 301 | 99 | |
| GenBank mRNA | BC135513.1 | - | 2 - 3 | 164 | 99 | |
| BC153334.1 | - | 2 - 3 | 301 | 99 | ||
More Information
Set Membership: |
|

cxcl14
Genomic Map