Assay Details
| Gene Symbol: rras | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114248.1 | NP_001107720.1 | 3 - 4 | 332 | 70 | |
| GenBank mRNA | BC158407.1 | - | 3 - 4 | 332 | 70 | |
More Information
Set Membership: |
|

rras
Genomic Map