Assay Details
| Gene Symbol: rnf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114261.2 | NP_001107733.2 | 2 - 3 | 216 | 77 | |
| GenBank mRNA | CR443913.1 | - | 2 - 3 | 214 | 77 | |
| CR443914.1 | - | 1 - 2 | 452 | 77 | ||
| EL642961.1 | - | 1 - 2 | 707 | 77 | ||
More Information
Set Membership: |
|

rnf7
Genomic Map