Assay Details
| Gene Symbol: eef1aknmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114492.1 | NP_001107964.1 | 3 - 4 | 1135 | 74 | |
| GenBank mRNA | BC159322.1 | - | 3 - 4 | 1135 | 74 | |
| CR942513.1 | - | 2 - 3 | 920 | 74 | ||
More Information
Set Membership: |
|

eef1aknmt
Genomic Map