Assay Details
| Gene Symbol: mbip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114493.1 | NP_001107965.1 | 2 - 3 | 277 | 60 | |
| GenBank mRNA | BC158957.1 | - | 2 - 3 | 277 | 60 | |
| CU024964.1 | - | 2 - 3 | 270 | 60 | ||
More Information
Set Membership: |
|

mbip
Genomic Map