Assay Details
| Gene Symbol: cdc25c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114496.2 | NP_001107968.1 | 13 - 14 | 1380 | 68 | |
| XM_012958406.2 | XP_012813860.1 | 13 - 14 | 1307 | 68 | ||
| XM_018091902.1 | XP_017947391.1 | 12 - 13 | 1277 | 68 | ||
| GenBank mRNA | BC159061.1 | - | 13 - 14 | 1381 | 68 | |
| CU075412.1 | - | 13 - 14 | 1413 | 68 | ||
More Information
Set Membership: |
|

cdc25c
Genomic Map