Assay Details
| Gene Symbol: dimt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122722.1 | NP_001116194.1 | 4 - 5 | 321 | 80 | |
| GenBank mRNA | BC123077.1 | - | 4 - 5 | 277 | 80 | |
| BC160444.1 | - | 4 - 5 | 321 | 80 | ||
| BC169134.1 | - | 4 - 5 | 324 | 80 | ||
| CU075813.1 | - | 4 - 5 | 282 | 80 | ||
More Information
Set Membership: |
|

dimt1
Genomic Map