Assay Details
| Gene Symbol: chn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122793.1 | NP_001116265.1 | 2 - 3 | 674 | 65 | |
| XM_012969930.2 | XP_012825384.1 | 8 - 9 | 1111 | 65 | ||
| GenBank mRNA | BC161070.1 | - | 2 - 3 | 674 | 65 | |
| CT030309.1 | - | 8 - 9 | 849 | 65 | ||
More Information
Set Membership: |
|

chn1
Genomic Map