Assay Details
| Gene Symbol: zfand4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012966840.1 | XP_012822294.1 | 2 - 3 | 470 | 63 | |
| XM_012966841.1 | XP_012822295.1 | 2 - 3 | 470 | 63 | ||
| GenBank mRNA | BC158157.1 | - | 2 - 3 | 414 | 63 | |
| CU075512.1 | - | 1 - 2 | 210 | 63 | ||
More Information
Set Membership: |
|

zfand4
Genomic Map