Assay Details
Gene Symbol: hnf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123448.1 | NP_001116920.1 | 8 - 9 | 1767 | 103 | |
GenBank mRNA | BC161562.1 | - | 8 - 9 | 1767 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
