Assay Details
| Gene Symbol: akt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123464.1 | NP_001116936.1 | 2 - 3 | 264 | 61 | |
| GenBank mRNA | BC161640.1 | - | 2 - 3 | 264 | 61 | |
More Information
Set Membership: |
|

akt2
Genomic Map