Assay Details
| Gene Symbol: dkk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123476.1 | NP_001116948.1 | 6 - 7 | 768 | 56 | |
| GenBank mRNA | BC161739.1 | - | 6 - 7 | 768 | 56 | |
More Information
Set Membership: |
|

dkk3
Genomic Map