Assay Details
| Gene Symbol: cyld | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123488.2 | NP_001116960.1 | 8 - 9 | 1839 | 71 | |
| XM_012960593.2 | XP_012816047.1 | 9 - 10 | 1985 | 71 | ||
| XM_012960594.2 | XP_012816048.1 | 9 - 10 | 1913 | 71 | ||
| XM_012960595.2 | XP_012816049.1 | 9 - 10 | 1986 | 71 | ||
| XM_018092917.1 | XP_017948406.1 | 9 - 10 | 2250 | 71 | ||
| GenBank mRNA | BC160532.1 | - | 8 - 9 | 1844 | 71 | |
More Information
Set Membership: |
|

cyld
Genomic Map