Assay Details
| Gene Symbol: pros1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126067.1 | NP_001119539.1 | 7 - 8 | 807 | 64 | |
| GenBank mRNA | BC166387.1 | - | 7 - 8 | 807 | 64 | |
More Information
Set Membership: |
|

pros1
Genomic Map