Assay Details
| Gene Symbol: slain2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126069.1 | NP_001119541.1 | 2 - 3 | 675 | 72 | |
| XM_012968579.2 | XP_012824033.1 | 2 - 3 | 989 | 72 | ||
| XM_012968580.1 | XP_012824034.1 | 2 - 3 | 718 | 72 | ||
| XM_012968581.2 | XP_012824035.1 | 2 - 3 | 794 | 72 | ||
| XM_018096299.1 | XP_017951788.1 | 2 - 3 | 742 | 72 | ||
| GenBank mRNA | BC121298.1 | - | 2 - 3 | 662 | 72 | |
| BC166063.1 | - | 2 - 3 | 675 | 72 | ||
More Information
Set Membership: |
|

slain2
Genomic Map