Assay Details
| Gene Symbol: gca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126075.1 | NP_001119547.1 | 6 - 7 | 520 | 65 | |
| GenBank mRNA | BC135414.1 | - | 6 - 7 | 518 | 65 | |
| BC166134.1 | - | 6 - 7 | 520 | 65 | ||
More Information
Set Membership: |
|

gca
Genomic Map