Assay Details
| Gene Symbol: opn1sw | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126076.1 | NP_001119548.1 | 2 - 3 | 521 | 96 | |
| GenBank mRNA | BC166308.1 | - | 2 - 3 | 521 | 96 | |
| CU025212.1 | - | 2 - 3 | 533 | 96 | ||
More Information
Set Membership: |
|

opn1sw
Genomic Map