Assay Details
| Gene Symbol: march9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126078.1 | NP_001119550.1 | 1 - 2 | 895 | 64 | |
| XM_012956375.2 | XP_012811829.1 | 2 - 3 | 1482 | 64 | ||
| GenBank mRNA | BC166307.1 | - | 1 - 2 | 895 | 64 | |
More Information
Set Membership: |
|

march9
Genomic Map