Assay Details
| Gene Symbol: march9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126078.1 | NP_001119550.1 | 2 - 3 | 1053 | 105 | |
| XM_012956375.2 | XP_012811829.1 | 3 - 4 | 1640 | 105 | ||
| GenBank mRNA | BC155516.1 | - | 1 - 2 | 172 | 105 | |
| BC166307.1 | - | 2 - 3 | 1053 | 105 | ||
More Information
Set Membership: |
|

march9
Genomic Map