Assay Details
| Gene Symbol: lcmt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126499.1 | NP_001119971.1 | 648 | 58 | ||
| GenBank mRNA | BC158186.1 | - | 648 | 58 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

lcmt2| Gene Symbol: lcmt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126499.1 | NP_001119971.1 | 648 | 58 | ||
| GenBank mRNA | BC158186.1 | - | 648 | 58 | ||
Set Membership: |
|