Assay Details
| Gene Symbol: cds2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126503.1 | NP_001119975.1 | 8 - 9 | 952 | 63 | |
| GenBank mRNA | BC158218.1 | - | 8 - 9 | 952 | 63 | |
| CR942673.2 | - | 1009 | 63 | |||
More Information
Set Membership: |
|

cds2
Genomic Map