Assay Details
| Gene Symbol: ces3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126547.1 | NP_001120019.1 | 6 - 7 | 859 | 98 | |
| GenBank mRNA | BC158318.1 | - | 6 - 7 | 859 | 98 | |
More Information
Set Membership: |
|

ces3
Genomic Map